2-Nitrotoluen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

N0275-25G N0275-500G
TCIAN0275-25GEA 132 DKK
TCIAN0275-25G TCIAN0275-500G
2-Nitrotoluen ≥99.0% (by GC)
2-Nitrotoluen
Formel: C₇H₇NO₂
Kogepunkt: 222°C
Smeltepunkt: –1 °C
Massefylde: 1,163 g/cm³ (20 °C)
Flammepunkt: 106°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007157
CAS-nummer: 88-72-2
EINECS: 201-853-3
UN nummer: 1664
ADR: 6.1,II
Merck Index: 14,06650
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ab2b5c6a25724906d3f4fa126045a2f7aaabdbb&vid=c2f26448c920c2d0a4b2be4e2f39e72d39890f1e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR