1-Ethyl-2-nitrobenzen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Ethylnitrobenzene

E0153-25G E0153-500G
TCIAE0153-25GEA 132 DKK
TCIAE0153-25G TCIAE0153-500G
1-Ethyl-2-nitrobenzen ≥99.0% (by GC)
1-Ethyl-2-nitrobenzen
Formel: C₈H₉NO₂
Kogepunkt: 232°C
Smeltepunkt: –11 °C
Massefylde: 1,1236 g/cm³ (20 °C)
MDL Number: MFCD00024294
CAS-nummer: 612-22-6
EINECS: 210-299-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a16fc096d537bd8cd35e95334092cdf232f1316b&vid=4228a3e19cefffc503d1f54e8b388c31567d14bd&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR