3-iodtoluen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Iodo-3-methylbenzene
I0216-25G
I0216-500G
TCIAI0216-25GEA
296
DKK
TCIAI0216-25G
TCIAI0216-500G
3-iodtoluen ≥98.0% (by GC)
3-iodtoluen
Formel:
C₇H₇I Kogepunkt: 213°C Smeltepunkt: -27°C Massefylde: 1,6835 g/cm³ (20 °C) Flammepunkt: 82°C |
MDL Number:
MFCD00001050 CAS-nummer: 625-95-6 EINECS: 210-918-5 |
|
Specifikationer Testresultater
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...