4-Nitro-α,α,α-trifluortoluen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Nitro-alpha,alpha,alpha-trifluorotoluene, 4-Nitrobenzotrifluoride, 1-Nitro-4-(trifluoromethyl)benzene

N0902-25G N0902-5G
TCIAN0902-25GEA 2720 DKK
TCIAN0902-25G TCIAN0902-5G
4-Nitro-α,α,α-trifluortoluen ≥98.0% (by GC)
4-Nitro-α,α,α-trifluortoluen
Formel: C₇H₄F₃NO₂
Kogepunkt: 82°C
Smeltepunkt: 40…41 °C
MDL Number: MFCD00007358
CAS-nummer: 402-54-0
EINECS: 206-948-3
UN nummer: 3431
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73f3878e73c074e02a51f5a26d348b812fbfa6fc&vid=e4c6e6159c6c09dd2606624da2d033a2eb90083e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to lump
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR