4-Fluortoluen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1-Fluoro-4-methylbenzene

F0041-25ML F0041-500ML
TCIAF0041-25MLEA 140 DKK
TCIAF0041-25ML TCIAF0041-500ML
4-Fluortoluen ≥99.0% (by GC)
4-Fluortoluen
Formel: C₇H₇F
Kogepunkt: 116°C
Smeltepunkt: -56°C
Massefylde: 1 g/cm³ (20 °C)
Flammepunkt: 10°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000358
CAS-nummer: 352-32-9
EINECS: 206-520-6
UN nummer: 2388
ADR: 3,II
Merck Index: 14,04180
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f502a0d6dc7f92e7324a10e4d424f7c45d0f18b&vid=08b042d4bbe60044a35bd454c05ce2d8d0600a9f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR