3-Fluorbenzylchlorid ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1-(Chloromethyl)-3-fluorobenzene, α-Chloro-m-fluorotoulene, 3-Fluorobenzyl chloride

F0210-25G F0210-5G
TCIAF0210-25GEA 559 DKK
TCIAF0210-25G TCIAF0210-5G
3-Fluorbenzylchlorid ≥95.0% (by GC)
3-Fluorbenzylchlorid
Formel: C₇H₆ClF
Kogepunkt: 180°C
Massefylde: 1,21 g/cm³ (20 °C)
Flammepunkt: 58°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000904
CAS-nummer: 456-42-8
EINECS: 207-264-8
UN nummer: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73b0a81c400b39f2962326a0ee1001cd8e39a0db&vid=7e794d642c99ea192ba29debd8af981a5cab9eac&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR