α,3-Dichlortoluen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
1-Chloro-3-(chloromethyl)benzene, 3-Chlorobenzyl chloride, 3-Chlorobenzylchloride
C0580-25G
C0580-100G
TCIAC0580-25GEA
279
DKK
TCIAC0580-25G
TCIAC0580-100G
α,3-Dichlortoluen ≥98.0% (by GC)
α,3-Dichlortoluen
Formel:
C₇H₆Cl₂ Kogepunkt: 222°C Smeltepunkt: 69…71 °C Massefylde: 1,27 g/cm³ (20 °C) Flammepunkt: 98°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000905 CAS-nummer: 620-20-2 EINECS: 210-629-4 UN nummer: 2235 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...