(4-Chlorbutyl)benzen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P1389-25G P1389-5G
TCIAP1389-25GEA 2610 DKK
TCIAP1389-25G TCIAP1389-5G
(4-Chlorbutyl)benzen ≥98.0% (by GC)
(4-Chlorbutyl)benzen
Formel: (C₆H₅)(CH₂)₄Cl
Flammepunkt: 104°C
MDL Number: MFCD00037117
CAS-nummer: 4830-93-7
EINECS: 225-408-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f7d239c8f86513af44766a6d710ff07402c111b&vid=3c79c670bcb297624e290d5983cbfe553232c79d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR