Heptanophenon ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H0369-500G H0369-25G
TCIAH0369-500GEA 3680 DKK
TCIAH0369-500G TCIAH0369-25G
Heptanophenon ≥98.0% (by GC)
Heptanophenon
Formel: C₆H₅C(O)(CH₂)₅CH₃
Kogepunkt: 283°C
Smeltepunkt: 17 °C
Massefylde: 0,9516 g/cm³ (20 °C)
MDL Number: MFCD00009539
CAS-nummer: 1671-75-6
EINECS: 216-802-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=170fad3b95bcce55b33995cb3831081610f9b65a&vid=261575f5f4456b54ecce144498d36801cb94e127&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR