4-Ethylbenzaldehyd ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: p-Ethylbenzaldehyde

E0325-25ML E0325-500ML E0325-100ML
TCIAE0325-25MLEA 206 DKK
TCIAE0325-25ML TCIAE0325-500ML TCIAE0325-100ML
4-Ethylbenzaldehyd ≥98.0% (by GC)
4-Ethylbenzaldehyde
Formel: C₉H₁₀O
Kogepunkt: 221°C
Massefylde: 0,97 g/cm³ (20 °C)
Flammepunkt: 92°C
MDL Number: MFCD00006956
CAS-nummer: 4748-78-1
EINECS: 225-268-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b4c143a2bc08e4f9eddf4acdbee6ec050f22841&vid=3b6403becf1672ae3dcc332ba79517d2bca7adb6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR