4-tert-Butylbenzaldehyd ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-(tert-Butyl)benzaldehyde
B1760-25G
B1760-500G
B1760-100G
TCIAB1760-25GEA
362
DKK
TCIAB1760-25G
TCIAB1760-500G
TCIAB1760-100G
4-tert-Butylbenzaldehyd ≥95.0% (by GC)
4-tert-Butylbenzaldehyd
Formel:
C₁₁H₁₄O Molekylvægt: 162.23 g/mol Kogepunkt: 246°C Flammepunkt: 101°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00035742 CAS-nummer: 939-97-9 EINECS: 213-367-9 UN nummer: 3082 ADR: 9 III |
|
Specifikationer Testresultater
Appearance | Colorless to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...