4-Brom-3-nitrotoluen ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Bromo-4-methyl-2-nitrobenzene
B1854-25G
B1854-5G
TCIAB1854-25GEA
706
DKK
TCIAB1854-25G
TCIAB1854-5G
4-Brom-3-nitrotoluen ≥96.0% (by GC)
4-Brom-3-nitrotoluen
Formel:
C₇H₆BrNO₂ Kogepunkt: 152°C Smeltepunkt: 35 °C Massefylde: 1,57 g/cm³ (20 °C) |
MDL Number:
MFCD00024180 CAS-nummer: 5326-34-1 EINECS: 226-203-6 |
|
Specifikationer Testresultater
Appearance | White or Colorles to Yellow to Orange powder to lump to clear liquid |
Purity (GC) | min. 96.0 % |
Freezing point | 29.0 to 33.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...