α,4-Dibrom-2-Fluortoluen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Bromo-1-(bromomethyl)-2-fluorobenzene, 4-Bromo-2-fluorobenzyl bromide

B1934-25G B1934-5G
TCIAB1934-25GEA 534 DKK
TCIAB1934-25G TCIAB1934-5G
α,4-Dibrom-2-Fluortoluen ≥98.0% (by GC)
α,4-Dibrom-2-Fluortoluen
Formel: C₇H₅Br₂F
Molekylvægt: 267.92 g/mol
Kogepunkt: 126
Smeltepunkt: 30
MDL Number: MFCD00055467
CAS-nummer: 76283-09-5
EINECS: 278-412-7
UN nummer: 2923
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5382a8aa24b7ece1ede510687807635ceecce64&vid=131da1f79309991a6200897ddc9b3a60e530b518&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystaline
Purity (GC) min. 98.0 %
Melting point 35.0 to 38.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR