2-Brom-α,α,α-trifluortoluen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Bromo-2-(trifluoromethyl)benzene, 2-Bromobenzotrifluoride

B0663-25G B0663-250G
TCIAB0663-25GEA 181 DKK
TCIAB0663-25G TCIAB0663-250G
2-Brom-α,α,α-trifluortoluen ≥98.0% (by GC)
2-Brom-α,α,α-trifluortoluen
Formel: C₇H₄BrF₃
Kogepunkt: 171°C
Massefylde: 1,651 g/cm³ (30 °C)
Flammepunkt: 51°C
MDL Number: MFCD00000373
CAS-nummer: 392-83-6
EINECS: 206-879-9
UN nummer: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6a61d364aa4d03a9585991c6490e2fa70cadbf0&vid=b167ddb81474dc5a6de66c55d399b83ce2c11c49&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR