α-Brom-4-Fluortoulen ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
α-Bromo-p-fluorotoulene, 4-Fluorobenzyl bromide, p-Fluorobenzyl bromide
F0214-25G
F0214-5G
TCIAF0214-25GEA
616
DKK
TCIAF0214-25G
TCIAF0214-5G
α-Brom-4-Fluortoulen ≥97.0% (by GC)
α-Brom-4-Fluortoulen
Formel:
C₇H₆BrF Kogepunkt: 177°C Massefylde: 1,517 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000359 CAS-nummer: 459-46-1 EINECS: 207-291-5 UN nummer: 3265 ADR: 8,II |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...