4-Brom-2-methylbenzoesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Bromo-2-methylbenzoic acid
B3337-25G
B3337-5G
TCIAB3337-25GEA
3940
DKK
TCIAB3337-25G
TCIAB3337-5G
4-Brom-2-methylbenzoesyre ≥98.0% (by GC, titration analysis)
4-Brom-2-methylbenzoesyre
Formel:
C₈H₇BrO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00040905 CAS-nummer: 68837-59-2 EINECS: 272-437-7 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 179.0 to 185.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...