4-Pyridinboronsyrepinacolester ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Pyridine-4-boronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
T2349-1G
T2349-5G
TCIAT2349-1GEA
238
DKK
TCIAT2349-1G
TCIAT2349-5G
4-Pyridinboronsyrepinacolester ≥98.0% (by GC, titration analysis)
4-Pyridinboronsyrepinacolester
Formel:
C₁₁H₁₆BNO₂ |
MDL Number:
MFCD01319051 CAS-nummer: 181219-01-2 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 150.0 to 154.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...