1-Benzyloxy-4-brombenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4-Benzyloxybromobenzene, 1-Bromo-4-(phenylmethoxy)benzene
B2127-25G
B2127-5G
TCIAB2127-25GEA
690
DKK
TCIAB2127-25G
TCIAB2127-5G
1-Benzyloxy-4-brombenzen ≥98.0% (by GC)
1-Benzyloxy-4-brombenzen
Formel:
C₁₃H₁₁BrO |
MDL Number:
MFCD00028016 CAS-nummer: 6793-92-6 |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 60.0 to 64.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...