1-Benzyloxy-3-brombenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
3-Benzyloxybromobenzene, 1-(Benzyloxy)-3-bromobenzene
B2307-25G
B2307-5G
TCIAB2307-25GEA
821
DKK
TCIAB2307-25G
TCIAB2307-5G
1-Benzyloxy-3-brombenzen ≥98.0% (by GC)
1-Benzyloxy-3-brombenzen
Formel:
C₁₃H₁₁BrO |
MDL Number:
MFCD00155065 CAS-nummer: 53087-13-1 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 60.0 to 64.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...