2-Brombenzylbromid ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Bromobenzyl bromide
D1514-25G
D1514-5G
TCIAD1514-25GEA
485
DKK
TCIAD1514-25G
TCIAD1514-5G
2-Brombenzylbromid ≥98.0% (by GC)
2-Brombenzylbromid
Formel:
C₇H₆Br₂ Molekylvægt: 249.93 g/mol Kogepunkt: 253 Smeltepunkt: 34 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000173 CAS-nummer: 3433-80-5 EINECS: 222-334-8 UN nummer: 3261 ADR: 8 III |
|
Specifikationer Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...