Methyl3-pentenoat ≥90.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
trans-3-Pentenoic acid methyll ester, Methyl trans-3-Pentenoate
P1210-25ML
P1210-5ML
TCIAP1210-25MLEA
2120
DKK
TCIAP1210-25ML
TCIAP1210-5ML
Methyl3-pentenoat ≥90.0% (by GC)
Methyl3-pentenoat
Formel:
C₆H₁₀O₂ Molekylvægt: 114.14 g/mol Kogepunkt: 138°C Flammepunkt: 24°C |
MDL Number:
MFCD00191576 CAS-nummer: 818-58-6 EINECS: 212-453-3 UN nummer: 3272 ADR: 3 III |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...