3-Phenylpropylbromid ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Bromo-3-phenylpropane

B1237-25G B1237-500G B1237-100G
TCIAB1237-25GEA 156 DKK
TCIAB1237-25G TCIAB1237-500G TCIAB1237-100G
3-Phenylpropylbromid ≥97.0% (by GC)
3-Phenylpropylbromid
Formel: BrH₂CCH₂CH₂(C₆H₅)
Molekylvægt: 199.09 g/mol
Kogepunkt: 129°C
Flammepunkt: 101°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000257
CAS-nummer: 637-59-2
EINECS: 211-294-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99cd433d5b916d0de80fa3f79d0af558ce61a3a5&vid=14911328f8d84ec81bb46aac3bdab3d0c57a3cd3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR