1-Phenyl-1-propyn ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Propynylbenzene, Prop-1-yn-1-ylbenzene

P0219-25ML P0219-5ML
TCIAP0219-25MLEA 1810 DKK
TCIAP0219-25ML TCIAP0219-5ML
1-Phenyl-1-propyn ≥97.0% (by GC)
1-Phenyl-1-propyn
Formel: C₆H₅CCCH₃
Kogepunkt: 181°C
Massefylde: 0,942 g/cm³ (15 °C)
Flammepunkt: 62°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00009272
CAS-nummer: 673-32-5
EINECS: 211-607-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d89d04285dfbdf6d84a07ecd892322418d12980&vid=b94b983bab456cfd67ac2d715b8e2217938801dd&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR