1,2,3-Trimethoxybenzen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T1102-25G T1102-500G
TCIAT1102-25GEA 206 DKK
TCIAT1102-25G TCIAT1102-500G
1,2,3-Trimethoxybenzen ≥99.0% (by GC)
1,2,3-Trimethoxybenzen
Formel: C₉H₁₂O₃
Kogepunkt: 235°C
Smeltepunkt: 43…45 °C
Massefylde: 1,1118 g/cm³ (45 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008358
CAS-nummer: 634-36-6
EINECS: 211-207-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5855476dccfd97dbb2079af9b946d864583a75cb&vid=97e8c4b4a51de32c73b01c1049fe0a0d04a5e54b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 43.0 to 46.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR