3,4-Dimethoxyphenol ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D3221-25G D3221-5G
TCIAD3221-25GEA 2180 DKK
TCIAD3221-25G TCIAD3221-5G
3,4-Dimethoxyphenol ≥97.0% (by GC)
3,4-Dimethoxyphenol
Formel: C₈H₁₀O₃
Kogepunkt: 167°C
Smeltepunkt: 83 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008390
CAS-nummer: 2033-89-8
EINECS: 217-995-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e62a28a996a756fa75e86447fed659ee33529f95&vid=0c63597d3b3eedeadedeef7635a10451e22ad43d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 79.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR