2,2'-Dipyridylamin ≥99.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2,2'-Iminodipyridine, Di(pyridin-2-yl)amine, Chlorpheniramine Related Compound B
D1200-25G
D1200-5G
TCIAD1200-25GEA
1850
DKK
TCIAD1200-25G
TCIAD1200-5G
2,2'-Dipyridylamin ≥99.0% (by GC, titration analysis)
2,2'-Dipyridylamin
Formel:
C₁₀H₉N₃ Molekylvægt: 171.2 g/mol Kogepunkt: 178°C |
MDL Number:
MFCD00006247 CAS-nummer: 1202-34-2 EINECS: 214-864-3 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 94.0 to 97.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...