1,2-Diethoxybenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
D0452-25G D0452-5G
TCIAD0452-25GEA 854 DKK
TCIAD0452-25G TCIAD0452-5G
1,2-Diethoxybenzen ≥98.0% (by GC)
1,2-Diethoxybenzen
Formel: C₁₀H₁₄O₂
Kogepunkt: 220°C
MDL Number: MFCD00015143
CAS-nummer: 2050-46-6
EINECS: 218-089-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f611d3806ac9e04aea3da3198989a3627381ded&vid=0f611d3806ac9e04aea3da3198989a3627381ded&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 45.0 deg-C
Solubility in Methanol almost transparency