1-Butyl-4-methylpyridiniumchlorid ≥99.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Butyl-4-methylpyridinium Chloride
B3426-25G
B3426-5G
TCIAB3426-25GEA
1990
DKK
TCIAB3426-25G
TCIAB3426-5G
1-Butyl-4-methylpyridiniumchlorid ≥99.0% (by HPLC, titration analysis)
1-Butyl-4-methylpyridinium chloride
Formel:
C₁₀H₁₆ClN |
MDL Number:
MFCD03095439 CAS-nummer: 112400-86-9 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 158.0 to 162.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...