Geraniol ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(E)-3,7-Dimethylocta-2,6-dien-1-ol
G0027-25ML
G0027-500ML
G0027-100ML
TCIAG0027-25MLEA
132
DKK
TCIAG0027-25ML
TCIAG0027-500ML
TCIAG0027-100ML
Geraniol ≥96.0% (by GC)
Geraniol
Formel:
C₁₀H₁₈O Molekylvægt: 154.25 g/mol Flammepunkt: 112°C |
MDL Number:
MFCD00002917 CAS-nummer: 106-24-1 EINECS: 203-377-1 Merck Index: 14,04403 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...