1-Methyl-4-piperidon ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-Methyl-4-piperidone

M0530-25G M0530-500G M0530-100G
TCIAM0530-25GEA 222 DKK
TCIAM0530-25G TCIAM0530-500G TCIAM0530-100G
1-Methyl-4-piperidon ≥98.0% (by GC, titration analysis)
1-Methyl-4-piperidon
Formel: C₆H₁₁NO
Kogepunkt: 72°C
Flammepunkt: 58°C
MDL Number: MFCD00006191
CAS-nummer: 1445-73-4
EINECS: 215-895-5
UN nummer: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc9c7f01590f7cab3042351c1232cacd97f2748e&vid=fc9c7f01590f7cab3042351c1232cacd97f2748e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR