1-Ethyl-4-piperidon ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
E0498-25ML
E0498-100ML
TCIAE0498-25MLEA
731
DKK
TCIAE0498-25ML
TCIAE0498-100ML
1-Ethyl-4-piperidon ≥98.0% (by GC, titration analysis)
1-Ethyl-4-piperidon
Formel:
C₇H₁₃NO Kogepunkt: 80°C Flammepunkt: 68°C |
MDL Number:
MFCD00013356 CAS-nummer: 3612-18-8 EINECS: 222-781-9 |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...