Guaiacol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Methoxyphenol
M0121-25G
M0121-500G
TCIAM0121-25GEA
156
DKK
TCIAM0121-25G
TCIAM0121-500G
Guaiacol ≥98.0% (by GC)
Guaiacol
Formel:
C₇H₈O₂ Kogepunkt: 205°C Smeltepunkt: 28…32 °C Massefylde: 1,1534 g/cm³ (0 °C) Flammepunkt: 82 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002185 CAS-nummer: 90-05-1 EINECS: 201-964-7 ADR: 6.1,III Merck Index: 14,04553 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...