Anisol ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methyl phenyl ether, Methoxybenzene

A0492-25G A0492-500G
TCIAA0492-25GEA 132 DKK
TCIAA0492-25G TCIAA0492-500G
Anisol ≥99.0% (by GC)
Anisol
Formel: C₇H₈O
Molekylvægt: 108.14 g/mol
Kogepunkt: 154°C
Smeltepunkt: -37
Flammepunkt: 51°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008354
CAS-nummer: 100-66-3
EINECS: 202-876-1
UN nummer: 2222
ADR: 3 III
Merck Index: 14,00669
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=893f34641defc879cc849f4f487f87a3de8bad7a&vid=287d79b8b8184c90e0a4221bd5ab5211863ac92b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR