N,N-Dimethyl-4-nitrosoanilin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4-Nitroso-N,N-dimethylaniline
N0262-25G
N0262-500G
TCIAN0262-25GEA
534
DKK
TCIAN0262-25G
TCIAN0262-500G
N,N-Dimethyl-4-nitrosoanilin ≥98.0% (by GC, titration analysis)
N,N-Dimethyl-4-nitrosoanilin
Formel:
C₈H₁₀N₂O Molekylvægt: 150.18 g/mol Kogepunkt: 259 Smeltepunkt: 87 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002063 CAS-nummer: 138-89-6 EINECS: 205-343-1 UN nummer: 1369 ADR: 4.2 II Merck Index: 14,06638 |
|
Specifikationer Testresultater
Appearance | Yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 84.0 to 86.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...