N,N-Dimethylanilin ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

D0665-25ML D0665-500ML
TCIAD0665-25MLEA 140 DKK
TCIAD0665-25ML TCIAD0665-500ML
N,N-Dimethylanilin ≥99.0% (by GC, titration analysis)
N,N-Dimethylanilin
Formel: C₈H₁₁N
Molekylvægt: 121.18 g/mol
Kogepunkt: 194°C
Smeltepunkt: 2.5
Flammepunkt: 73°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008304
CAS-nummer: 121-69-7
EINECS: 204-493-5
UN nummer: 2253
ADR: 6.1 II
Merck Index: 14,03234
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee6f5d1672df6000ac049d36f3a72950fc4ae98a&vid=28230f904d9e056152b1ac5cf4f0ccf8eed62fa0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR