3-iodanisol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

I0379-25G I0379-5G
TCIAI0379-25GEA 567 DKK
TCIAI0379-25G TCIAI0379-5G
3-iodanisol ≥98.0% (by GC)
3-iodanisol
Formel: C₇H₇IO
Kogepunkt: 245°C
Massefylde: 1,765 g/cm³ (20 °C)
MDL Number: MFCD00001048
CAS-nummer: 766-85-8
EINECS: 212-173-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=592f529101319160f9f86cc3fc573c06ce5d2e0d&vid=592f529101319160f9f86cc3fc573c06ce5d2e0d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR