2-Iodanisol ≥98.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Stabilization:
Stabilised with copper chip
I0382-25G
I0382-100G
TCIAI0382-25GEA
362
DKK
TCIAI0382-25G
TCIAI0382-100G
2-Iodanisol ≥98.0% (by GC) stabilised
2-Iodanisol
Formel:
C₇H₇IO Kogepunkt: 238°C Massefylde: 1,445 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001039 CAS-nummer: 529-28-2 EINECS: 208-456-4 Merck Index: 14,05028 |
|
Specifikationer Testresultater
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...