3-Iodanilin ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

I0047-25G I0047-5G
TCIAI0047-25GEA 772 DKK
TCIAI0047-25G TCIAI0047-5G
3-Iodanilin ≥99.0% (by GC, titration analysis)
3-Iodanilin
Formel: C₆H₆IN
Molekylvægt: 219.02 g/mol
Kogepunkt: 116°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00007781
CAS-nummer: 626-01-7
EINECS: 210-922-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c1dd413512c4d08189d9af66f9d61a4a0d5e8ed&vid=36bd64ea2633e9060f885099ca28d57990d2b29e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to lump
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR