1,3-Diiodbenzen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D0607-25G D0607-5G
TCIAD0607-25GEA 706 DKK
TCIAD0607-25G TCIAD0607-5G
1,3-Diiodbenzen ≥99.0% (by GC)
1,3-Diiodbenzen
Formel: C₆H₄I₂
Molekylvægt: 329.91 g/mol
Kogepunkt: 284°C
Smeltepunkt: 39
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00041731
CAS-nummer: 626-00-6
EINECS: 210-921-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46c198e565a5ec29dcf65662b5c01d40fa2da447&vid=32fd1c00be8f5143169fe0164792d9f805a637cc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 33.0 to 37.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR