4-Fluorthiophenol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Fluorobenzene-1-thiol, 1-Fluoro-4-mercaptobenzene, 4-Fluorobenzenethiol, p-Fluorothiophenol

F0393-25G F0393-5G
TCIAF0393-25GEA 534 DKK
TCIAF0393-25G TCIAF0393-5G
4-Fluorthiophenol ≥98.0% (by GC)
4-Fluorthiophenol
Formel: C₆H₅FS
Molekylvægt: 128.17 g/mol
Kogepunkt: 168°C
Flammepunkt: 54°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004846
CAS-nummer: 371-42-6
EINECS: 206-737-6
UN nummer: 3336
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed4e25dd361e8d927b9c821bd4502bd2ed25f575&vid=8d543a7e35fa793a09f86814aeff400948dd70c2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR