1-Fluor-2,4-dinitrobenzen ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
D0835-25G
D0835-500G
TCIAD0835-25GEA
279
DKK
TCIAD0835-25G
TCIAD0835-500G
1-Fluor-2,4-dinitrobenzen ≥99.0% (by GC)
1-Fluor-2,4-dinitrobenzen
Formel:
C₆H₃FN₂O₄ Kogepunkt: 296°C Smeltepunkt: 24…26 °C Massefylde: 1,48 g/cm³ (20 °C) Flammepunkt: ≥ 100 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007056 CAS-nummer: 70-34-8 EINECS: 200-734-3 UN nummer: 2811 ADR: 6.1,II Merck Index: 14,04172 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Yellow to Green powder to lump to clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...