1,2,3-Trifluor-4-nitrobenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2,3,4-Trifluoronitrobenzene
T1539-25G
T1539-5G
TCIAT1539-25GEA
460
DKK
TCIAT1539-25G
TCIAT1539-5G
1,2,3-Trifluor-4-nitrobenzen ≥98.0% (by GC)
1,2,3-Trifluor-4-nitrobenzen
Formel:
C₆H₂F₃NO₂ Molekylvægt: 177.08 g/mol Kogepunkt: 92°C Flammepunkt: 93°C |
MDL Number:
MFCD00041546 CAS-nummer: 771-69-7 EINECS: 212-238-4 |
|
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...