3-Fluoranilin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
F0101-25G
F0101-250G
TCIAF0101-25GEA
288
DKK
TCIAF0101-25G
TCIAF0101-250G
3-Fluoranilin ≥98.0% (by GC, titration analysis)
3-Fluoranilin
Formel:
C₆H₆FN Molekylvægt: 111.12 g/mol Kogepunkt: 186°C Smeltepunkt: -2 Flammepunkt: 77°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007758 CAS-nummer: 372-19-0 EINECS: 206-747-0 UN nummer: 2941 ADR: 6.1 III |
|
Specifikationer Testresultater
Appearance | Colorless to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...