2-Fluoranilin ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

F0032-25G F0032-500G F0032-100G
TCIAF0032-25GEA 132 DKK
TCIAF0032-25G TCIAF0032-500G TCIAF0032-100G
2-Fluoranilin ≥99.0% (by GC)
2-Fluoranilin
Formel: C₆H₆FN
Molekylvægt: 111.12 g/mol
Kogepunkt: 170°C
Smeltepunkt: -29
Flammepunkt: 60°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007642
CAS-nummer: 348-54-9
EINECS: 206-478-9
UN nummer: 2941
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4faf2f8e350c0b8b5d2bff52ba64a5276d6ef9ef&vid=7af8da641e09a4cfdb8216257c9262488581ad8c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR