3,5-Difluorbenzoesyre ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D2596-25G
D2596-5G
TCIAD2596-25GEA
1320
DKK
TCIAD2596-25G
TCIAD2596-5G
3,5-Difluorbenzoesyre ≥98.0% (by titrimetric analysis)
3,5-Difluorbenzoesyre
Formel:
C₇H₄F₂O₂ Kogepunkt: 242 °C (1013 hPa) Smeltepunkt: 122 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010323 CAS-nummer: 455-40-3 |
|
Specifikationer Testresultater
Appearance | White to Amber to Dark purple powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 121.0 to 124.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...