4-Fluorbenzaldehyd ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

F0191-25ML F0191-250ML
TCIAF0191-25MLEA 140 DKK
TCIAF0191-25ML TCIAF0191-250ML
4-Fluorbenzaldehyd ≥98.0% (by GC)
4-Fluorobenzaldehyde
Formel: C₇H₅FO
Kogepunkt: 181°C
Smeltepunkt: –10 °C
Massefylde: 1,15 g/cm³ (20 °C)
Flammepunkt: 56°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003378
CAS-nummer: 459-57-4
EINECS: 207-293-6
UN nummer: 1989
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3ad4879cca545b0fefe2450c720747d5452dea5&vid=50c64be518a1ece4be32ce0ba69aa98db7b7fb00&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR