3,4-Difluorbenzaldehyd ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D2340-25G
D2340-5G
TCIAD2340-25GEA
1060
DKK
TCIAD2340-25G
TCIAD2340-5G
3,4-Difluorbenzaldehyd ≥97.0% (by GC)
3,4-Difluorobenzaldehyde
Formel:
C₇H₄F₂O Kogepunkt: 53 °C (1013 hPa) Massefylde: 1,29 g/cm³ (20 °C) Flammepunkt: 66°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010328 CAS-nummer: 34036-07-2 EINECS: 422-180-3 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...