3,4,5-Trifluorbenzaldehyd ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T2321-1G T2321-5G
TCIAT2321-1GEA 271 DKK
TCIAT2321-1G TCIAT2321-5G
3,4,5-Trifluorbenzaldehyd ≥98.0% (by GC)
3,4,5-Trifluorobenzaldehyde
Formel: C₇H₃F₃O
Kogepunkt: 176 - 183 °C (1013 hPa)
Smeltepunkt: 25 °C
Massefylde: 1,42 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00083523
CAS-nummer: 132123-54-7
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62c202462fded24c5229fde27afae8db82ec9030&vid=23161aa106efbcd1f87e09d5b678df400afa6598&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR