4-Chlorphenol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: p-Chlorophenol

C0243-25G C0243-500G
TCIAC0243-25GEA 140 DKK
TCIAC0243-25G TCIAC0243-500G
4-Chlorphenol ≥98.0% (by GC, titration analysis)
4-Chlorphenol
Formel: C₆H₅ClO
Molekylvægt: 128.56 g/mol
Kogepunkt: 220°C
Flammepunkt: 121
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002318
CAS-nummer: 106-48-9
EINECS: 203-402-6
UN nummer: 2020
ADR: 6.1 III
Merck Index: 14,02154
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdb9bdb02c6f516ae80f260df108812bf6b4c618&vid=754a177b52fecdf7254f94608714b74142533e05&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR