2,5-Dichloranisol

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1,4-Dichloro-2-methoxybenzene

D2605-25G D2605-5G
TCIAD2605-25GEA 1740 DKK
TCIAD2605-25G TCIAD2605-5G
2,5-Dichloranisol
2,5-Dichloranisol
Formel: C₇H₆Cl₂O
Kogepunkt: 225 - 227 °C (1013 hPa)
Smeltepunkt: 24 °C
Massefylde: 1,333 g/cm³ (20 °C)
MDL Number: MFCD00061123
CAS-nummer: 1984-58-3
EINECS: 217-852-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01bdc537c3b662144c2e44a478b849225e3b452c&vid=517f7cc5ad113ccc2ce43a8fe89c1ccf252c86eb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR